Benzyl 2-(diethoxyphosphoryl)acetate 95%

Supplier: Ambeed
Warning

A243733-10G A243733-250MG A243733-1G A243733-25G A243733-5G
103811-542EA 12 USD
103811-542 103851-368 76826-798 76826-800 77304-282 77304-284 77304-286
Benzyl 2-(diethoxyphosphoryl)acetate 95%
Benzyl 2-(diethoxyphosphoryl)acetate

Precaución: for Research Use Only

Formula: C₁₃H₁₉O₅P
MW: 286.26 g/mol
Boiling Pt: 386.9 °C (760 mm Hg)
Flash Pt: 201.3 °C
Storage Temperature: Ambient
MDL Number: MFCD09263511
CAS Number: 7396-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR