Benzyl (6-hydroxyhexyl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: 6-(N-Benzyloxycarbonylamino)-1-hexanol

A184280-100G A184280-10G A184280-250MG A184280-1G A184280-25G A184280-5G
103815-736EA 10 USD
103815-736 103846-298 76769-660 76769-662 76769-664 77083-906 77083-908 77083-910
Benzyl (6-hydroxyhexyl)carbamate 97%
Benzyl (6-hydroxyhexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₃
MW: 251.32 g/mol
Boiling Pt: 419.6 °C (760 mm Hg)
Melting Pt: 80…82 °C
Flash Pt: 207.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00859036
CAS Number: 17996-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR