Benzyl(4-oxocyclohexyl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: Benzyl (4-oxocyclohexyl)carbamate, 4-N-Cbz-aminocyclohexanone

A158455-10G A158455-250MG A158455-1G A158455-25G A158455-5G
103805-132EA 6 USD
103805-132 103843-342 77011-094 77011-096 77319-204 77319-206 77319-208
Benzyl(4-oxocyclohexyl)carbamate 97%
Benzyl(4-oxocyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
Boiling Pt: 427.3 °C (760 mm Hg)
Melting Pt: 86…87 °C
Storage Temperature: Ambient
MDL Number: MFCD06658393
CAS Number: 16801-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR