Benzyl (3-oxocyclohexyl)carbamate 98%

Supplier: Ambeed
Warning

A141836-100MG A141836-10G A141836-250MG A141836-1G A141836-25G A141836-5G
103803-068EA 15 USD
103803-068 77285-776 77285-778 77285-780 77299-252 77299-254 77299-256
Benzyl (3-oxocyclohexyl)carbamate 98%
Benzyl (3-oxocyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061944
CAS Number: 320590-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR