Benzyl(3-oxocyclobutyl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: 3-Aminocyclobutanone, N-CBZ protected, Benzyl (3-oxocyclobutyl)carbamate

A160137-100G A160137-10G A160137-250MG A160137-5G A160137-1G A160137-25G
103805-340EA 24 USD
103805-340 103843-570 77028-590 77028-592 77028-594 77300-442 77319-490 77319-492
Benzyl(3-oxocyclobutyl)carbamate 97%
Benzyl(3-oxocyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃
MW: 219.2400 g/mol
Boiling Pt: 403.3 °C (760 mm Hg)
Flash Pt: 197.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13152267
CAS Number: 130369-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR