Benzyl (3-bromopropyl)carbamate 97%

Supplier: Ambeed
Warning

A212382-10G A212382-250MG A212382-1G A212382-5G
103818-194EA 63 USD
103818-194 103848-892 76999-646 76999-648 77087-360 77087-362
Benzyl (3-bromopropyl)carbamate 97%
Benzyl (3-bromopropyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO₂
MW: 272.14 g/mol
Boiling Pt: 366.4 °C (760 mm Hg)
Melting Pt: 37…39 °C
Flash Pt: 175.4 °C
Storage Temperature: Freezer
MDL Number: MFCD01737871
CAS Number: 39945-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR