Benzyl (2-oxo-1,2-dihydropyridin-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A119568-100MG A119568-10G A119568-250MG A119568-1G A119568-5G
103799-784EA 83 USD
103799-784 76810-398 76810-400 76810-402 77311-902 77311-904
Benzyl (2-oxo-1,2-dihydropyridin-3-yl)carbamate 95%
Benzyl (2-oxo-1,2-dihydropyridin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O₃
MW: 244.25 g/mol
Boiling Pt: 474.3 °C (760 mm Hg)
Flash Pt: 240.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07772020
CAS Number: 147269-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR