2-Bromoethylamine, N-CBZ protected 98%

Supplier: Ambeed
Warning

A111992-10G A111992-250MG A111992-25G A111992-1G A111992-5G A111992-100G
103797-934EA 6 USD
103797-934 103823-760 76995-268 76995-270 76995-272 77310-568 77310-570 77880-901
2-Bromoethylamine, N-CBZ protected 98%
2-Bromoethylamine, N-CBZ protected

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂
Boiling Pt: 361.1 °C (760 mm Hg)
Melting Pt: 38…41 °C
Flash Pt: 172.2 °C
Storage Temperature: Freezer
MDL Number: MFCD09909490
CAS Number: 53844-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR