Benzyl(2-aminoethyl)(methyl)carbamate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzyl (2-aminoethyl)(methyl)carbamate hydrochloride
A278789-250MG
A278789-1G
103813-428EA
84
USD
103813-428
103853-372
76816-334
77093-296
Benzyl(2-aminoethyl)(methyl)carbamate hydrochloride 97%
Benzyl(2-aminoethyl)(methyl)carbamate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₇ClN₂O₂ MW: 244.72 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11112220 CAS Number: 162576-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...