Benzothiazole-5-carboxylic acid 98%

Supplier: Ambeed
Warning

A149351-250MG A149351-1G A149351-5G A149351-100MG
103804-032EA 56 USD
103804-032 103842-150 76768-090 77299-726 77299-728 77567-118
Benzothiazole-5-carboxylic acid 98%
Benzothiazole-5-carboxylic acid

Precaución: For research use only.

Formula: C₈H₅NO₂S
MW: 179.2 g/mol
Boiling Pt: 387.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06796380
CAS Number: 68867-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR