Benzhydrazide 98%

Supplier: Ambeed
Warning

Synonyms: Benzoylhydrazine, Benzoic acid hydrazide, Benzohydrazide, Benzoic hydrazide

A898931-100G A898931-10G A898931-500G A898931-5G A898931-1000G A898931-25G
103810-598EA 107 USD
103810-598 76698-586 76698-588 76698-590 76698-592 76845-608 76845-610
Benzhydrazide 98%
Benzhydrazide

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 112…114 °C
Flash Pt: 139.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00007596
CAS Number: 613-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR