Benzofuran-7-carboxylic acid 95%

Supplier: Ambeed
Warning

A106998-100MG A106998-250MG A106998-1G
103797-110EA 47 USD
103797-110 103822-868 76687-268 76687-270 77283-532
Benzofuran-7-carboxylic acid 95%
Benzofuran-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆O₃
Boiling Pt: 325.559 °C (760 mm Hg)
Melting Pt: 166…170 °C
Flash Pt: 150.693 °C
Storage Temperature: Ambient
MDL Number: MFCD10000617
CAS Number: 90484-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR