Benzofuran-6-carboxylic acid 98%

Supplier: Ambeed
Warning

A255852-100MG A255852-10G A255852-250MG A255852-1G A255852-25G A255852-5G
103812-218EA 14 USD
103812-218 103852-078 76692-620 76692-622 76692-624 77091-544 77091-546 77091-548
Benzofuran-6-carboxylic acid 98%
Benzofuran-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆O₃
MW: 162.142 g/mol
Boiling Pt: 325.6 °C (760 mm Hg)
Flash Pt: 150.693 °C
Storage Temperature: Ambient
MDL Number: MFCD01006751
CAS Number: 77095-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR