Benzofuran-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A146937-250MG A146937-1G
103803-722EA 21 USD
103803-722 103841-818 77259-066 77317-174
Benzofuran-5-carboxylic acid 97%
Benzofuran-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆O₃
MW: 162.1400 g/mol
Boiling Pt: 325.6 °C (760 mm Hg)
Melting Pt: 189…190 °C
Flash Pt: 150.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01006742
CAS Number: 90721-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR