Benzofuran-5-carbaldehyde 98%

Supplier: Ambeed
Warning

A251240-1G A251240-5G A251240-250MG
103811-976EA 50 USD
103811-976 103851-818 77091-196 77091-198 77261-996
Benzofuran-5-carbaldehyde 98%
Benzofuran-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₆O₂
Boiling Pt: 251.5 °C (760 mm Hg)
Melting Pt: 50.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411182
CAS Number: 10035-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR