Benzo[b]furan-3-boronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzofuran-3-ylboronic acid
A109076-100MG
A109076-250MG
A109076-25G
A109076-1G
A109076-10G
103797-442EA
24
USD
103797-442
103823-230
76869-626
76869-628
76869-630
77283-806
77670-544
Benzo[b]furan-3-boronic acid 98%
Benzo[b]furan-3-boronic acid
Precaución: for Research Use Only
Formula:
C₈H₇BO₃ Storage Temperature: Freezer |
MDL Number:
MFCD06801687 CAS Number: 317830-83-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...