Benzofuran-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A287325-1G A287325-5G A287325-100MG
103813-746EA 119 USD
103813-746 103834-888 76837-430 76837-432 77194-244
Benzofuran-3-carboxylic acid 95%
Benzofuran-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆O₃
Boiling Pt: 325.6 °C (760 mm Hg)
Melting Pt: 162 °C
Flash Pt: 150.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01006722
CAS Number: 26537-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR