Benzofuran-3-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: Benzofuran-3-carbaldehyde

A187646-100MG A187646-10G A187646-250MG A187646-1G A187646-25G A187646-5G
103816-030EA 26 USD
103816-030 103846-620 76784-814 76784-816 76784-818 77084-322 77084-324 77084-326
Benzofuran-3-carboxaldehyde 95%
Benzofuran-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₆O₂
MW: 146.143 g/mol
Melting Pt: 39 °C
Flash Pt: 110.192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03412541
CAS Number: 4687-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR