3-Coumaranone 97%

Supplier: Ambeed
Warning

Synonyms: Benzofuran-3(2H)-one

A173791-1G A173791-25G A173791-5G A173791-100G A173791-10G A173791-500G
103806-870EA 20 USD
103806-870 103845-170 77082-448 77082-450 77082-452 77236-360 77236-362 77746-147
3-Coumaranone 97%
3-Coumaranone

Precaución: for Research Use Only

Formula: C₈H₆O₂
MW: 134.13 g/mol
Melting Pt: 101…103 °C
Storage Temperature: Freezer
MDL Number: MFCD00051810
CAS Number: 7169-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR