Benzofuran-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A315071-25G A315071-5G A315071-100G A315071-10G A315071-1G A315071-500G
103825-056EA 6 USD
103825-056 103835-982 76838-810 76838-812 77263-266 77263-268 77306-124 77748-615
Benzofuran-2-carboxylic acid 98%
Benzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆O₃
Melting Pt: 193…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00005848
CAS Number: 496-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR