Benzo[ghi]perylene 95%

Supplier: Ambeed
Warning

A194228-100MG A194228-250MG A194228-1G A194228-5G A194228-10G
103816-634EA 47 USD
103816-634 103847-246 77192-090 77192-092 77302-398 77302-400 77567-816
Benzo[ghi]perylene 95%
Benzo[ghi]perylene

Precaución: For research use only.

Formula: C₂₂H₁₂
MW: 276.34 g/mol
Boiling Pt: >500 °C (760 mm Hg)
Melting Pt: 277…279 °C
Storage Temperature: Ambient
MDL Number: MFCD00004135
CAS Number: 191-24-2
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR