Benzo[d]thiazole-7-carboxylic acid 96%

Supplier: Ambeed
Warning

A416612-1G A416612-100MG A416612-250MG
103827-318EA 67 USD
103827-318 103838-364 76841-948 76979-224 77882-093
Benzo[d]thiazole-7-carboxylic acid 96%
Benzo[d]thiazole-7-carboxylic acid

Precaución: For research use only

Formula: C₈H₅NO₂S
Boiling Pt: 387.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD13195419
CAS Number: 677304-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR