1,3-benzoxazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: Benzo[d]oxazole-5-carboxylic acid

A193091-100MG A193091-250MG A193091-5G
103816-498EA 25 USD
103816-498 103847-114 76999-050 76999-052 77085-012
1,3-benzoxazole-5-carboxylic acid 95%
1,3-benzoxazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₃
MW: 163.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09954950
CAS Number: 15112-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR