1,3-Benzoxazol-6-amine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Aminobenzoxazole, Benzo[d]oxazol-6-amine

A183848-1G A183848-25G A183848-5G A183848-250MG
103815-678EA 28 USD
103815-678 103846-240 77083-844 77083-846 77083-848 77191-774
1,3-Benzoxazol-6-amine 97%
1,3-Benzoxazol-6-amine

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.10 g/mol
Melting Pt: 88 °C
Flash Pt: 124.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03791187
CAS Number: 177492-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR