1,3-Benzoxazol-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: Benzo[d]oxazol-2-amine, 2-Aminobenzoxazole

A345780-1G A345780-25G A345780-5G A345780-100G A345780-10G A345780-250MG
103825-932EA 6 USD
103825-932 103836-916 76839-982 76839-984 76839-986 77142-150 77142-152 77142-154
1,3-Benzoxazol-2-amine 98%
1,3-Benzoxazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.1353 g/mol
Boiling Pt: 278 °C (760 mm Hg)
Melting Pt: 128…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00182850
CAS Number: 4570-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR