Benzo[d]isoxazol-5-amine 98%

Supplier: Ambeed
Warning

A137066-250MG A137066-1G A137066-5G
103802-398EA 98 USD
103802-398 76972-388 77298-882 77315-340
Benzo[d]isoxazol-5-amine 98%
Benzo[d]isoxazol-5-amine

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13190363
CAS Number: 239097-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR