3,4-(Methylenedioxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-(Methylenedioxy)benzeneboronic acid, Benzo[d][1,3]dioxol-5-ylboronic acid

A124476-100G A124476-250MG A124476-25G A124476-1G A124476-5G A124476-10G
103800-536EA 6 USD
103800-536 77149-348 77149-350 77149-352 77312-672 77312-674 77571-470
3,4-(Methylenedioxy)phenylboronic acid 98%
3,4-(Methylenedioxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BO₄
Boiling Pt: 347.3±52.0 °C (760 mm Hg)
Melting Pt: 224…229 °C
Flash Pt: 163.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01009695
CAS Number: 94839-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR