2H-1,3-benzodioxol-4-ol 98%

Supplier: Ambeed
Warning

Synonyms: Benzo[d][1,3]dioxol-4-ol

A166956-100MG A166956-250MG A166956-1G
103806-112EA 85 USD
103806-112 77163-374 77163-376 77300-860
2H-1,3-benzodioxol-4-ol 98%
2H-1,3-benzodioxol-4-ol

Precaución: Inert atmosphere, room temperature

Formula: C₇H₆O₃
MW: 138.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092232
CAS Number: 69393-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR