2,1,3-Benzothiadiazol-5-ylamine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2,1,3-benzothiadiazole, Benzo[c][1,2,5]thiadiazol-5-amine

A253844-1G A253844-25G A253844-5G A253844-250MG
103812-114EA 28 USD
103812-114 103851-966 77091-398 77091-400 77091-402 77249-992
2,1,3-Benzothiadiazol-5-ylamine 97%
2,1,3-Benzothiadiazol-5-ylamine

Precaución: for Research Use Only

Formula: C₆H₅N₃S
MW: 151.1900 g/mol
Boiling Pt: 303 °C (760 mm Hg)
Melting Pt: 112…115 °C
Flash Pt: 137.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00185826
CAS Number: 874-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR