Benzofurazan 97%

Supplier: Ambeed
Warning

A120288-100G A120288-25G A120288-1G A120288-5G
103799-910EA 31 USD
103799-910 76688-078 76688-080 77312-014 77312-016
Benzofurazan 97%
Benzofurazan

Precaución: for Research Use Only

Formula: C₆H₄N₂O
Boiling Pt: 186 °C (760 mm Hg)
Melting Pt: 47…51 °C
Flash Pt: 70.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00068064
CAS Number: 273-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR