Benzo[b]thiophene-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A181596-1G A181596-5G
103807-626EA 103 USD
103807-626 103846-002 77301-728 77301-730
Benzo[b]thiophene-4-carboxylic acid 98%
Benzo[b]thiophene-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆O₂S
MW: 178.208 g/mol
Melting Pt: 188…189 °C
Storage Temperature: Freezer
MDL Number: MFCD01929340
CAS Number: 10134-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR