Benzo[b]thiophene-3(2H)-one 1,1-dioxide 97%

Supplier: Ambeed
Warning

A564221-25G A564221-250MG A564221-10G
103828-764EA 15 USD
103828-764 103839-902 76844-016 77292-560 77617-392
Benzo[b]thiophene-3(2H)-one 1,1-dioxide 97%
Benzo[b]thiophene-3(2H)-one 1,1-dioxide

Precaución: for Research Use Only

Formula: C₈H₆O₃S
Boiling Pt: 424.6 °C (760 mm Hg)
Melting Pt: 131.1…135.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00159164
CAS Number: 1127-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR