Benzo[b]thiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A137128-10G A137128-500G A137128-1G A137128-25G A137128-5G A137128-100G
103802-414EA 5 USD
103802-414 77258-704 77258-706 77315-364 77315-366 77315-368 77747-970
Benzo[b]thiophene-2-carboxylic acid 98%
Benzo[b]thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆O₂S
Boiling Pt: 376.2 °C (760 mm Hg)
Melting Pt: 238…243 °C
Flash Pt: 181.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00051636
CAS Number: 6314-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR