Benzo[b]thiophene-2-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Benzo[b]thiophene-2-carbaldehyde

A137891-10G A137891-1G A137891-25G A137891-5G
103802-524EA 14 USD
103802-524 76688-884 77315-514 77315-516 77315-518
Benzo[b]thiophene-2-carboxaldehyde 98%
Benzo[b]thiophene-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₆OS
MW: 162.21 g/mol
Boiling Pt: 303.2 °C (760 mm Hg)
Melting Pt: 30…32 °C
Storage Temperature: Ambient
MDL Number: MFCD01075041
CAS Number: 3541-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR