Benzo[b]thiophen-5-ol 97%

Supplier: Ambeed
Warning

A147097-100MG A147097-250MG A147097-1G
103803-734EA 186 USD
103803-734 103841-830 76984-494 76984-496 77299-566
Benzo[b]thiophen-5-ol 97%
Benzo[b]thiophen-5-ol

Precaución: for Research Use Only

Formula: C₈H₆OS
Storage Temperature: Refrigerator
MDL Number: MFCD16876937
CAS Number: 19301-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR