Benzo[b]thiophene-2-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Benzo[b]thiophen-2-ylboronic acid, 2-Benzothienylboronic acid

A261855-25G A261855-5G A261855-10G A261855-100G
103812-542EA 9 USD
103812-542 103852-432 77092-042 77092-044 77238-524 77304-834 77746-438
Benzo[b]thiophene-2-boronic acid 98%
Benzo[b]thiophene-2-boronic acid

Precaución: for Research Use Only

Formula: C₈H₇BO₂S
Boiling Pt: 390.2 °C (760 mm Hg)
Melting Pt: 254…256 °C
Flash Pt: 189.8 °C
Storage Temperature: Freezer
MDL Number: MFCD01075674
CAS Number: 98437-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR