Benzofuran-2-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Benzo[b]furan-2-carboxaldehyde, Benzofuran-2-carbaldehyde

A207838-1G A207838-25G A207838-5G A207838-100G A207838-10G
103817-832EA 18 USD
103817-832 103848-518 77086-808 77086-810 77086-812 77260-980 77260-982
Benzofuran-2-carboxaldehyde 98%
Benzofuran-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₆O₂
MW: 146.145 g/mol
Boiling Pt: 251.5 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00015463
CAS Number: 4265-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR