Benzo[b]benzo[4,5]thieno[2,3-d]thiophene 97%
Supplier: Ambeed
|
Warning
|
A259960-100MG
A259960-10G
A259960-25G
A259960-1G
103812-472EA
21
USD
103812-472
103852-354
76827-110
76827-112
76827-114
77091-932
Benzo[b]benzo[4,5]thieno[2,3-d]thiophene 97%
Benzo[b]benzo[4,5]thieno[2,3-d]thiophene
Precaución: for Research Use Only
Formula:
C₁₄H₈S₂ Storage Temperature: Ambient |
MDL Number:
MFCD02114004 CAS Number: 248-70-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...