1,3,5-Benzenetrimethanol 96%

Supplier: Ambeed
Warning

Synonyms: 1,3,5-Tris(hydroxymethyl)benzene, Benzene-1,3,5-triyltrimethanol

A354880-1G A354880-5G A354880-25G
103826-134EA 26 USD
103826-134 103837-138 76840-266 76840-268 77195-256
1,3,5-Benzenetrimethanol 96%
1,3,5-Benzenetrimethanol

Precaución: for Research Use Only

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Melting Pt: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD01108484
CAS Number: 4464-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR