1,3,5-Triformylbenzene 96%

Supplier: Ambeed
Warning

Synonyms: 1,3,5-benzenetricarbaldehyde, Trimesic formaldehyde, Benzene-1,3,5-tricarbaldehyde

A372528-1G A372528-5G A372528-100MG A372528-10G A372528-250MG
103799-632EA 28 USD
103799-632 103826-506 103837-524 76840-784 76840-786 77195-500 77195-502 77195-504
1,3,5-Triformylbenzene 96%
1,3,5-Triformylbenzene

Precaución: for Research Use Only

Formula: C₉H₆O₃
Melting Pt: 156…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00182470
CAS Number: 3163-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR