Benzene-1,3,5-triamine trihydrochloride 95% (by HPLC)
Supplier: Ambeed
|
Warning
|
A804054-100MG
A804054-250MG
A804054-5G
A804054-1G
103841-070EA
418
USD
103841-070
76820-566
76820-568
76820-570
76845-318
Benzene-1,3,5-triamine trihydrochloride 95% (by HPLC)
Benzene-1,3,5-triamine trihydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₂Cl₃N₃ Boiling Pt: 440.9 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD00035496 CAS Number: 638-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...