1,2,4-Trihydroxybenzene 95%

Supplier: Ambeed
Danger

Synonyms: Benzene-1,2,4-triol, 1,2,4-Benzenetriol

A118069-10G A118069-25G A118069-5G
103798-984EA 8 USD
103798-984 103824-876 77271-340 77271-342 77311-666
1,2,4-Trihydroxybenzene 95%
1,2,4-Trihydroxybenzene

Precaución: for Research Use Only

Formula: C₆H₆O₃
MW: 126.11 g/mol
Melting Pt: 140 °C
Flash Pt: 34.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00002198
CAS Number: 533-73-3
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR