(R)-rel-3-((R)-1-Benzylpiperidin-3-yl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A214046-1MG A214046-25MG A214046-100MG A214046-250MG A214046-5G
103792-418EA 6 USD
103792-418 103799-492 103818-360 76691-582 77303-320 77877-668 77877-669 77877-670
(R)-rel-3-((R)-1-Benzylpiperidin-3-yl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride 98%
(R)-rel-3-((R)-1-Benzylpiperidin-3-yl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₂₈H₃₂ClN₃O₆
Storage Temperature: Freezer
MDL Number: MFCD06407650
CAS Number: 91599-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR