1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,2',2'',2'''-(((Ethane-1,2-diylbis(oxy))bis(2,1-phenylene))bis(azanetriyl))tetraacetic acid, BAPTA

A235410-25MG A235410-1MG A235410-250MG
103811-010EA 62 USD
103811-010 103850-778 76692-146 77626-246 77626-248
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₄N₂O₁₀
MW: 476.44 g/mol
Boiling Pt: 766.6 °C (760 mm Hg)
Melting Pt: 174 °C
Flash Pt: 417.4 °C
Storage Temperature: Freezer
MDL Number: MFCD00036255
CAS Number: 85233-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR