2-Nitroimidazole 99%

Supplier: Ambeed
Danger

Synonyms: Azomycin, 2-Nitro-1H-imidazole

A259140-10G A259140-1MG A259140-1G A259140-25G A259140-5G A259140-100G
103812-426EA 10 USD
103812-426 103852-302 76976-876 76976-878 77091-838 77091-840 77091-842 77672-308
2-Nitroimidazole 99%
2-Nitroimidazole

Precaución: for Research Use Only

Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: 287 °C
Flash Pt: 179.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00005185
CAS Number: 527-73-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR