Azetidine-2-carboxylic acid 98%

Supplier: Ambeed
Danger

Synonyms: azetidine-2-carboxylic acid

A217569-5G A217569-10G A217569-250MG A217569-50MG A217569-1G A217569-25G
103818-632EA 15 USD
103818-632 103849-360 77087-988 77097-866 77097-868 77097-870 77303-454 77303-456
Azetidine-2-carboxylic acid 98%
Azetidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₇NO₂
Melting Pt: 210…214 °C
Flash Pt: 106.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00066660
CAS Number: 2517-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR