Azetidine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Azetidine HCl

A129037-10G A129037-1G A129037-25G A129037-5G A129037-100G
103801-220EA 6 USD
103801-220 76971-964 77313-580 77313-582 77313-584 77670-826
Azetidine hydrochloride 95%
Azetidine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₈ClN
MW: 93.56 g/mol
Melting Pt: >300 °C
Flash Pt: 152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191762
CAS Number: 36520-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR