Azetidin-3-ol hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxyazetidine hydrochloride

A102357-100G A102357-10G A102357-25G A102357-500G A102357-5G
103822-054EA 5 USD
103822-054 76970-222 76970-224 76970-226 76970-228 77308-938
Azetidin-3-ol hydrochloride 97%
Azetidin-3-ol hydrochloride

Precaución: for Research Use Only

Formula: C₃H₈ClNO
Boiling Pt: 170.7 °C (760 mm Hg)
Melting Pt: 90…92 °C
Flash Pt: 120.4 °C
Storage Temperature: Ambient
MDL Number: MFCD02683887
CAS Number: 18621-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR