Azathioprine 98%

Supplier: Ambeed
Warning

Synonyms: 6-((1-Methyl-4-nitro-1H-imidazol-5-yl)thio)-9H-purine, 6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine

A101947-100MG A101947-1MG A101947-1G A101947-5G A101947-250MG A101947-25G
103796-216EA 41 USD
103796-216 103821-910 76687-058 76687-060 77282-868 77282-870 77599-492 77623-830
Azathioprine 98%
Azathioprine

Precaución: for Research Use Only

Formula: C₉H₇N₇O₂S
MW: 277.26 g/mol
Boiling Pt: 555.8 °C (760 mm Hg)
Melting Pt: 243…244 °C
Flash Pt: 290 °C
Storage Temperature: Freezer
MDL Number: MFCD00069203
CAS Number: 446-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR