Atropic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Phenylacrylic Acid, Ipratropium bromide Impurity D

A508395-1G A508395-25G A508395-5G A508395-100G A508395-500G
103828-356EA 14 USD
103828-356 103839-456 76843-438 76843-440 76843-442 77103-068 77103-070
Atropic acid 98%
Atropic acid

Precaución: for Research Use Only

Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 267.8 °C (760 mm Hg)
Flash Pt: 174.521 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00046531
CAS Number: 492-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR